Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGGT1B Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9548R

Specifications
PGGT1B | |
Polyclonal | |
Unconjugated | |
PGGT1B | |
2010207C17Rik; 2610100E13; AI451237; AI551093; BGG1; BGGI; CTC-428G20.3; Geranylgeranyl transferase type I subunit beta; Geranylgeranyl transferase type-1 subunit beta; geranylgeranyltransferase type 1 beta; geranylgeranyltransferase type I (GGTase-I); geranylgeranyltransferase type I beta subunit; GGT1; GGTase-I-beta; GGTI; Pggt1b; protein geranylgeranyltransferase type I subunit beta; protein geranylgeranyltransferase type I, beta subunit; Type I protein geranyl-geranyltransferase subunit beta | |
Rabbit | |
Protein A | |
RUO | |
5229 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P53609 | |
PGGT1B | |
KLH conjugated synthetic peptide derived from human PGGT1B/FNTB. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction