Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGGT1B Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2854670.1ML
Description
PGGT1B Polyclonal specifically detects PGGT1B in Rat samples. It is validated for Western Blot.Specifications
PGGT1B | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
5229 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
BGGI, EC 2.5.1, EC 2.5.1.59, Geranylgeranyl transferase type I subunit beta, geranylgeranyl transferase type-1 subunit beta, geranylgeranyltransferase type I beta subunit, GGTase-I-beta, GGTI, protein geranylgeranyltransferase type I, beta subunit, Type I protein geranyl-geranyltransferase subunit beta | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Rat PGGT1B | |
0.1 mL | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction