Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGK1 Recombinant Rabbit Monoclonal Antibody (4B3Q3)
GREENER_CHOICE

Catalog No. PIMA535173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535173 Supplier Invitrogen™ Supplier No. MA535173
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MSLSNKLTLD KLDVKGKRVV MRVDFNVPMK NNQITNNQRI KAAVPSIKFC LDNGAKSVVL MSHLGRPDGV PMPDKYSLEP VAVELKSLLG KDVLFLKDCV.

Also known as ATP:3-phosphoglycerate 1-phosphotransferase showed that the plasmin reductase isolated from conditioned medium of fibrosarcoma cells is the glycolytic enzyme phosphoglycerate kinase. They concluded that phosphoglycerate kinase not only functions in glycolysis but is secreted by tumor cells and participates in the angiogenic process as a disulfide reductase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGK1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4B3Q3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PGK1
Gene Accession No. P00558, P09411, P16617
Gene Alias cell migration-inducing gene 10 protein; epididymis secretory sperm binding protein Li 68p; HEL-S-68p; MIG10; OK/SW-cl.110; Pgk; Pgk1; Pgk-1; PGKA; Phosphoglycerate kinase; phosphoglycerate kinase 1; Primer recognition protein 2; PRP 2; YCR012W; YCR12W
Gene Symbols PGK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PGK1 (P00558).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18655, 24644, 5230
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.