Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGK1/PGK2 Recombinant Rabbit Monoclonal Antibody (HL1775)
GREENER_CHOICE

Catalog No. PIMA547177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547177 Supplier Invitrogen™ Supplier No. MA547177
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The protein encoded by this gene is a glycolytic enzyme that catalyzes the conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate The encoded protein may also act as a cofactor for polymerase alpha Additionally, this protein is secreted by tumor cells where it participates in angiogenesis by functioning to reduce disulfide bonds in the serine protease, plasmin, which consequently leads to the release of the tumor blood vessel inhibitor angiostatin The encoded protein has been identified as a moonlighting protein based on its ability to perform mechanistically distinct functions Deficiency of the enzyme is associated with a wide range of clinical phenotypes hemolytic anemia and neurological impairment Pseudogenes of this gene have been defined on chromosomes 19, 21 and the X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGK1/PGK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL1775
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Accession No. P29452, P29466, P43527, P48729, P97633, Q8BK63, Q8N752
Gene Alias HEL-S-68p; MIG10; PGKA; phosphoglycerate kinase 1
Host Species Rabbit
Immunogen Recombinant protein of human PGK1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 0, 113927, 122011, 12362, 1452, 834
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.