Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579815
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579815 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579815 Supplier Invitrogen™ Supplier No. PA579815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, rat brain tissue. IHC: Human Lung Cancer tissue.

Also known as ATP:3-phosphoglycerate 1-phosphotransferase showed that the plasmin reductase isolated from conditioned medium of fibrosarcoma cells is the glycolytic enzyme phosphoglycerate kinase. They concluded that phosphoglycerate kinase not only functions in glycolysis but is secreted by tumor cells and participates in the angiogenic process as a disulfide reductase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene PGK1
Gene Accession No. P00558, P09411, P16617
Gene Alias cell migration-inducing gene 10 protein; epididymis secretory sperm binding protein Li 68p; HEL-S-68p; MIG10; OK/SW-cl.110; Pgk; Pgk1; Pgk-1; PGKA; Phosphoglycerate kinase; phosphoglycerate kinase 1; Primer recognition protein 2; PRP 2; YCR012W; YCR12W
Gene Symbols PGK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human PGK1 (166-180aa FGTAHRAHSSMVGVN).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18655, 24644, 5230
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.