Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGK1 Rabbit anti-Human, Mouse, Unconjugated, Clone: EAB-16, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20316
Description
PGK1 Monoclonal antibody specifically detects PGK1 in Human, Mouse samples. It is validated for ICC, WB-Ce, IF, Flow Cyt.Specifications
PGK1 | |
Monoclonal | |
Unconjugated | |
PGK1 | |
Rabbit | |
100 μg | |
5230 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunocytochemistry, Western Blot, Immunofluorescence, Flow Cytometry | |
EAB-16 | |
phosphoglycerate kinase 1 | |
MGC117307, MGC142128, MGC8947, MIG10, PGKA | |
A synthetic peptide corresponding to human PGK1 | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction