Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGK2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299497F
Description
PGK2 Polyclonal antibody specifically detects PGK2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
PGK2 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
dJ417L20.2, EC 2.7.2.3, PGK-2, PGKB, PGKPS, phosphoglycerate kinase 2, phosphoglycerate kinase autosomal pseudogene, Phosphoglycerate kinase, testis specific | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PGK2. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
5232 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction