Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588885 Supplier Invitrogen™ Supplier No. PA588885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YTFLKVLNNM EIGASLFDEE GAKIVKDIMA KAQKNGVRIT FPVDFVTGDK FDENAQVGKA TVASGISPGW MGLDCGPESN KNHAQVVAQA RLIVWNGPLG VFEWDAFAKG TKALMDEIV; Positive Samples: U-87MG, Jurkat, HeLa, HepG2, Mouse testis, Mouse brain, Mouse heart, Rat testis, Rat brain; Cellular Location: Cytoplasm.

PGK2 is a form of phosphoglycerate kinase unique to spermatozoa. The autosomal PGK2 gene lacks introns and contains characteristics of a processed gene. The conservation of function in this processed PGK2 gene and its tissue-specific expression in spermatogenesis suggests that it exists as a compensatory response to the inactivation of the X-linked PGK1 gene in spermatogenic cells before meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGK2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PGK2
Gene Accession No. P07205, P09041
Gene Alias dJ417L20.2; epididymis secretory protein Li 272; HEL-S-272; Pgk2; Pgk-2; PGKB; PGKPS; phosphoglycerate kinase 2; phosphoglycerate kinase autosomal pseudogene; phosphoglycerate kinase, testis specific; Tcp-2; testicular tissue protein Li 139
Gene Symbols PGK2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 242-360 of human PGK2 (NP_6200612).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18663, 316265, 5232
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.