Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGL2 Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | PGL2 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PGL2 Polyclonal specifically detects PGL2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
PGL2 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
54949 | |
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human SDHAF2. The immunogen is located within the first 50 amino acids of SDHAF2. Amino Acid Squence: SFRRFYRGDSPTDSQKD | |
Primary | |
SDHAF2 antibody is predicted to not cross-react with other SDHAF protein family members. |
1 mg/mL | |
Polyclonal | |
Chicken | |
Human, Mouse, Rat | |
C11orf79, FLJ20487, hSDH5, paraganglioma or familial glomus tumors 2, PGL2chromosome 11 open reading frame 79, SDH5SDH assembly factor 2, succinate dehydrogenase assembly factor 2, mitochondrial, succinate dehydrogenase complex assembly factor 2, Succinate dehydrogenase subunit 5, mitochondrial | |
SDHAF2 | |
IgY | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title