Learn More
Promega pGL4.53[luc2/PGK] Vector
Catalog No. pre5011
Click to view available options
Quantity:
20 μg
Vectors are used as controls in dual-luciferase assays where Renilla or NanoLuc luciferase is the experimental reporter.
  • Vectors That Are Co-transfected into Cells as Controls in Dual-Reporter Assays
  • Co-transfect into cells where Renilla or NanoLuc(R) luciferase is the experimental reporter gene
  • SV40, PGK and TK promoter options offer different expression levels suitable for most dual-reporter assays
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.