Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGLS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531678
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531678 Supplier Invitrogen™ Supplier No. PA531678
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH. Predicted reactivity: Mouse (88%), Rat (88%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PGLS belongs to the glucosamine/galactosamine-6-phosphate isomerase family, 6-phosphogluconolactonase subfamily. It is implicated in the hydrolysis of 6-phosphogluconolactone to 6-phosphogluconate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGLS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PGLS
Gene Accession No. O95336, Q9CQ60
Gene Alias 1110030K05Rik; 6PGL; 6PGL {ECO:0000250; 6-phosphogluconolactonase; 6-phosphogluconolactonase {ECO:0000250; AI447866; epididymis secretory protein Li 304; HEL-S-304; hypothetical protein LOC616120; Pgls; pgls {ECO:0000250; Plgs; UniProtKB:O95336}
Gene Symbols PGLS
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PGLS. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25796, 66171
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.