Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGLS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544901
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544901 Supplier Invitrogen™ Supplier No. PA544901
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AAVLKRILED QEENPLPAAL VQPHTGKLCW FLDEAAARLL TVPFEKHSTL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rat: 93%.

PGLS belongs to the glucosamine/galactosamine-6-phosphate isomerase family, 6-phosphogluconolactonase subfamily. It is implicated in the hydrolysis of 6-phosphogluconolactone to 6-phosphogluconate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGLS
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PGLS
Gene Accession No. O95336
Gene Alias 1110030K05Rik; 6PGL; 6PGL {ECO:0000250; 6-phosphogluconolactonase; 6-phosphogluconolactonase {ECO:0000250; AI447866; epididymis secretory protein Li 304; HEL-S-304; hypothetical protein LOC616120; Pgls; pgls {ECO:0000250; Plgs; UniProtKB:O95336}
Gene Symbols PGLS
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PGLS.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25796
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.