Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGLYRP3/PGRPIA Antibody (187C434) - Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233249
Description
PGLYRP3/PGRPIA Monoclonal specifically detects PGLYRP3/PGRPIA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
PGLYRP3/PGRPIA | |
Monoclonal | |
Unconjugated | |
Q96LB9 | |
PGLYRP3 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 178-193 of human PGRP-1a. | |
0.1 mL | |
Primary | |
PGRP-1a (187C434) | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgM |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
187C434 | |
Western Blot 1:500-1:1000, Flow Cytometry 1ul/1 million cells, Immunocytochemistry/Immunofluorescence reported in scientific literature (Uehara et al (2005)), Flow (Cell Surface) reported in scientific literature (Uehara et al (2005)) | |
peptidoglycan recognition protein 3, peptidoglycan recognition protein I alpha, Peptidoglycan recognition protein I-alpha, Peptidoglycan recognition protein intermediate alpha, peptidoglycan recognition protein-I-alpha, PGLYRPIalphaMGC149197, PGRP-Ialpha, PGRPIAPGRP-I-alpha | |
Mouse | |
Unpurified | |
RUO | |
114771 | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction