Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGLYRP4/PGRP-I beta Antibody (186C426), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056721H
Description
PGLYRP4/PGRP-I beta Monoclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot.Specifications
PGLYRP4/PGRP-I beta | |
Monoclonal | |
HRP | |
Q96LB8 | |
PGLYRP4 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to amino acids 95-110 of human PGRP-1 beta. | |
0.1 mL | |
Primary | |
PGRP-1b (186C426) | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
186C426 | |
Western Blot | |
peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, PGRP-I-beta | |
Mouse | |
Protein G purified | |
RUO | |
57115 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction