Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGM1 Antibody (84G2), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259454C
This item is not returnable.
View return policy
Description
PGM1 Monoclonal antibody specifically detects PGM1 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
PGM1 | |
Monoclonal | |
DyLight 650 | |
EC 5.4.2, EC 5.4.2.2, Glucose phosphomutase 1, GSD14, PGM 1, phosphoglucomutase 1, phosphoglucomutase-1 | |
Recombinant human PGM1 (1-562aa) purified from E. coli | |
0.1 mL | |
Lipid and Metabolism | |
5236 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence | |
84G2 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction