Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119338
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119338 200 μL
PIPA5121688 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119338 Supplier Invitrogen™ Supplier No. PA5119338
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PGM1 Polyclonal Antibody for Western Blot, IHC (P)

Phosphoglucomutases (PGM; EC 5.4.2.2) catalyze the transfer of phosphate between the 1 and 6 positions of glucose. Isozymes of PGM are monomeric, with molecular masses of about 60 kD, and are encoded by several genes, including PGM1. In most cell types, PGM1 isozymes predominate, representing about 90% of total PGM activity. One exception is red cells, where PGM2 (MIM 172000) is a major isozyme (Putt et al., 1993 [PubMed 8257433]). [supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PGM1
Gene Accession No. P36871
Gene Alias 3230402E02Rik; Aciculin; CDG1T; Glucose phosphomutase 1; glucose phosphomutase 2; GSD14; PGM; PGM 1; PGM 2; Pgm1; Pgm-1; Pgm2; PGM5; PGMRP; PGM-RP; Phosphodeoxyribomutase; phosphoglucomutase 1; phosphoglucomutase 2; phosphoglucomutase 5; phosphoglucomutase isoform 2; phosphoglucomutase isoform1; phosphoglucomutase-1; Phosphoglucomutase-2; phosphoglucomutase-like protein 5; Phosphoglucomutase-related protein; phosphopentomutase; zgc:63718
Gene Symbols PGM1
Host Species Rabbit
Immunogen Recombinant protein Phosphoglucomutase 1. The antigen corresponds to amino acid range 1-235 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397566, 5236
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.