Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGM2L1 Monoclonal Antibody (OTI1C2), TrueMAB™, OriGene
SDP

Catalog No. 502543796
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3796 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3796 Supplier OriGene Supplier No. CF812133
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PGM2L1 Monoclonal antibody specifically detects PGM2L1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PGM2L1
Applications Western Blot
Classification Monoclonal
Clone OTI1C2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Hspa1l
Gene Accession No. P16627, P34931, P55063
Gene Alias 4931406N15Rik; AI553438; BM32A; FLJ32029; Glucose 1,6-bisphosphate synthase; im:7140576; im:7140576 protein; Pgm2l1; phosphoglucomutase 2 like 1; phosphoglucomutase 2-like 1; phosphoglucomutase-2-like 1; PMMLP; zgc:198285
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 400-641 of human HSPA1L (NP_005518) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15482, 24963, 3305
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.