Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGM2L1 Monoclonal Antibody (OTI5F5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812137


Description
PGM2L1 Monoclonal antibody specifically detects PGM2L1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
PGM2L1 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q6PCE3, Q8CAA7 | |
Mouse | |
Protein A/G | |
RUO | |
283209, 685076, 70974 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5F5 | |
Unconjugated | |
PGM2L1 | |
4931406N15Rik; AI553438; BM32A; FLJ32029; Glucose 1,6-bisphosphate synthase; im:7140576; im:7140576 protein; Pgm2l1; phosphoglucomutase 2 like 1; phosphoglucomutase 2-like 1; phosphoglucomutase-2-like 1; PMMLP; zgc:198285 | |
Full length human recombinant protein of human PGM2L1 (NP_775853) produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction