Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGM2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598545
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598545 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598545 Supplier Invitrogen™ Supplier No. PA598545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGM2L1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

PGM2L1 is the Glucose 1, 6-bisphosphate synthase using 1, 3-bisphosphoglycerate as a phosphate donor and a series of 1-phosphate sugars as acceptors, including glucose 1-phosphate, mannose 1-phosphate, ribose 1-phosphate and deoxyribose 1-phosphate. 5 or 6-phosphosugars are bad substrates, with the exception of glucose 6-phosphate. PGM2L1 also synthesizes ribose 1, 5-bisphosphate. PGM2L1 has only low phosphopentomutase and phosphoglucomutase activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGM2L1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PGM2L1
Gene Accession No. Q6PCE3
Gene Alias 4931406N15Rik; AI553438; BM32A; FLJ32029; Glucose 1,6-bisphosphate synthase; im:7140576; im:7140576 protein; Pgm2l1; phosphoglucomutase 2 like 1; phosphoglucomutase 2-like 1; phosphoglucomutase-2-like 1; PMMLP; zgc:198285
Gene Symbols PGM2L1
Host Species Rabbit
Immunogen Recombinant Human Glucose 1,6-bisphosphate synthase protein (101-400AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283209
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.