Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGM3 Antibody (OTI1A11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273362F
Description
PGM3 Monoclonal specifically detects PGM3 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
PGM3 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
PGM3 | |
Full length recombinant protein of human PGM3 (NP_056414) produced in HKE293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
IgG1 |
Flow Cytometry, Immunohistochemistry | |
OTI1A11 | |
Flow Cytometry, Immunohistochemistry | |
AGM1PAGMAcetylglucosamine phosphomutase, DKFZP434B187, EC 5.4.2, EC 5.4.2.3, FLJ11614, FLJ13623, N-acetylglucosamine-phosphate mutase, N-acetylglucosamine-phosphate mutase 1, PGM 3, phosphoacetylglucosamine mutase, phosphoglucomutase 3, Phosphoglucomutase-3 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
5238.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction