Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114087 Supplier Invitrogen™ Supplier No. PA5114087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVVQTAYANG SSTRYLEEVM KVPVYCTKTG VKHLHHKAQE FDIGVYFEAN For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 93%; Zebrafish: 93%.

Catalyzes the conversion of GlcNAc-6-P into GlcNAc-1-P during the synthesis of uridine diphosphate/UDP-GlcNAc, a sugar nucleotide critical to multiple glycosylation pathways including protein N- and O-glycosylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGM3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Pgm3
Gene Accession No. O95394
Gene Alias 2810473H05Rik; acetylglucosamine phosphomutase; Agm1; BB187688; C77933; GlcNAc-P mutase; IMD23; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Pgm3; Pgm-3; Phosphoacetylglucosamine mutase; phosphoglucomutase 3; phosphoglucomutase-3
Gene Symbols Pgm3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PGM3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5238
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.