Learn More
PGM3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304555AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Catalyzes the conversion of GlcNAc-6-P into GlcNAc-1-P during the synthesis of uridine diphosphate/UDP-GlcNAc, a sugar nucleotide critical to multiple glycosylation pathways including protein N- and O-glycosylation.Specifications
PGM3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95394 | |
PGM3 | |
Between 492 and 542 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PGM3 | |
AGM1, PAGM, PGM 3, N-acetylglucosamine-phosphate mutase 1, acetylglucosamine phosphomutase, phosphoacetylglucosamine mutase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5238 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.