Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGM5 (Aciculin) Mouse anti-Bovine, Rabbit, Mouse, Rat, Human, Unconjugated, Clone: 14F8 , MilliporeSigma™

Catalog No. MABT1503100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT1503-100 100 μg
MABT1503-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT1503-100 Supplier MilliporeSigma Supplier No. MABT1503100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PGM5 (Aciculin), clone 14F8, Cat. No. MABT1503, is a mouse monoclonal antibody that detects Phosphoglucomutase-like protein 5 and has been tested for use in ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGM5 (Aciculin)
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 14F8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Aciculin;Phosphoglucomutase-like Protein 5;PGM-RP
Gene Symbols PGM5;PGMRP
Host Species Mouse
Immunogen Cytoskeletal fraction of the human myometrium tissue extract.
Molecular Weight of Antigen ∽57 kDa observed; 62.22 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_068800
Test Specificity Clone 14F8 detects Phosphoglucomutase-like protein 5 in mouse and rat cardiac tissue and is predicted to react with human.
Target Species Bovine, Rabbit, Mouse, Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.