Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526852
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526852 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526852 Supplier Invitrogen™ Supplier No. PA526852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGP Polyclonal Antibody for Western Blot, IHC (P), Flow

Glycerol-3-phosphate phosphatase hydrolyzing glycerol-3-phosphate into glycerol. Thereby, regulates the cellular levels of glycerol-3-phosphate a metabolic intermediate of glucose, lipid and energy metabolism. Was also shown to have a 2-phosphoglycolate phosphatase activity and a tyrosine-protein phosphatase activity. However, their physiological relevance is unclear. In vitro, has also a phosphatase activity toward ADP, ATP, GDP and GTP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGP
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Pgp
Gene Accession No. A6NDG6
Gene Alias 1700012G19Rik; AI481330; Aspartate-based ubiquitous Mg(2+)-dependent phosphatase; AUM; G3PP; Glycerol-3-phosphate phosphatase; PGP; PGPase; phosphoglycolate phosphatase; RGD1307773
Gene Symbols Pgp
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 33-62 amino acids from the N-terminal region of human PGP.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283871
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.