Learn More
Invitrogen™ PGP Polyclonal Antibody

Description
Immunogen sequence: VVVGFDPHFS YMKLTKALRY LQQPGCLLVG TNMDNRLPLE NGRFIAGTGC LVRAVEMAAQ RQADIIGKPS RFIFDCVSQE YGINP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Specifications
Specifications
| Antigen | PGP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Pgp |
| Gene Accession No. | A6NDG6 |
| Gene Alias | 1700012G19Rik; AI481330; Aspartate-based ubiquitous Mg(2+)-dependent phosphatase; AUM; G3PP; Glycerol-3-phosphate phosphatase; PGP; PGPase; phosphoglycolate phosphatase; RGD1307773 |
| Gene Symbols | Pgp |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.