Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PGP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115748
Description
Antibody detects endogenous levels of total PGP.
Glycerol-3-phosphate phosphatase hydrolyzing glycerol-3-phosphate into glycerol. Thereby, regulates the cellular levels of glycerol-3-phosphate a metabolic intermediate of glucose, lipid and energy metabolism. Was also shown to have a 2-phosphoglycolate phosphatase activity and a tyrosine-protein phosphatase activity. However, their physiological relevance is unclear. In vitro, has also a phosphatase activity toward ADP, ATP, GDP and GTP.
Specifications
| PGP | |
| Polyclonal | |
| Unconjugated | |
| Pgp | |
| 1700012G19Rik; AI481330; Aspartate-based ubiquitous Mg(2+)-dependent phosphatase; AUM; G3PP; Glycerol-3-phosphate phosphatase; PGP; PGPase; phosphoglycolate phosphatase; RGD1307773 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 283871, 287115, 67078 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| A6NDG6, D3ZDK7, Q8CHP8 | |
| Pgp | |
| A synthesized peptide derived from human PGP(Accession A6NDG6), corresponding to amino acid residues F176-A226. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction