Learn More
Invitrogen™ PGP9.5 Monoclonal Antibody (BH7)

Description
Suggested positive control: bovine brain whole cell homogenate, SHSY-5Y or HEK 293 cell extract, and rat spinal cord/peripheral nerve homogenate, antigen standard for UCHL1 (transient overexpression lysate).
Specifications
Specifications
| Antigen | PGP9.5 |
| Applications | Immunohistochemistry (Free Floating), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | BH7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.035% sodium azide |
| Gene | Uchl1 |
| Gene Accession No. | P09936, P23356, Q00981, Q6SEG5, Q9GM50, Q9R0P9 |
| Gene Alias | AW822034; C88048; epididymis luminal protein 117; epididymis secretory protein Li 53; gad; gracile axonal dystrophy; HEL-117; HEL-S-53; MGC133697 protein; NDGOA; neuron cytoplasmic protein 9.5; OTTHUMP00000218137; OTTHUMP00000218139; OTTHUMP00000218140; OTTHUMP00000218141; PARK5; PGP 9.5; PGP9.5; PGP95; protein gene product 9.5; R75593; ubiquitin carboxyl-terminal esterase; ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase); ubiquitin carboxyl-terminal hydrolase isozyme L1; ubiquitin carboxyl-terminal hydrolase L1; ubiquitin carboxy-terminal hydrolase L1; ubiquitin C-terminal hydrolase; ubiquitin C-terminal hydrolase 1; ubiquitin C-terminal hydrolase L1; ubiquitin thioesterase L1; ubiquitin thiolesterase; ubiquitin thiolesterase L1; UCHL; UCHL1; UCH-L1; UCHL-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.