Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGP9.5 (UCHL1) Monoclonal Antibody (OTI5G2), TrueMAB™, OriGene
SDP

Catalog No. 502543417
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3417 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3417 Supplier OriGene Supplier No. CF814140
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PGP9.5 (UCHL1) Monoclonal antibody specifically detects PGP9.5 (UCHL1) in Human samples. It is validated for ELISA

Specifications

Antigen PGP9.5 (UCHL1)
Applications ELISA
Classification Monoclonal
Clone OTI5G2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Uchl1
Gene Accession No. P09936
Gene Alias AW822034; C88048; epididymis luminal protein 117; epididymis secretory protein Li 53; gad; gracile axonal dystrophy; HEL-117; HEL-S-53; MGC133697 protein; NDGOA; neuron cytoplasmic protein 9.5; OTTHUMP00000218137; OTTHUMP00000218139; OTTHUMP00000218140; OTTHUMP00000218141; PARK5; PGP 9.5; PGP9.5; PGP95; protein gene product 9.5; R75593; ubiquitin carboxyl-terminal esterase; ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase); ubiquitin carboxyl-terminal hydrolase isozyme L1; ubiquitin carboxyl-terminal hydrolase L1; ubiquitin carboxy-terminal hydrolase L1; ubiquitin C-terminal hydrolase; ubiquitin C-terminal hydrolase 1; ubiquitin C-terminal hydrolase L1; ubiquitin thioesterase L1; ubiquitin thiolesterase; ubiquitin thiolesterase L1; UCHL; UCHL1; UCH-L1; UCHL-1
Host Species Mouse
Immunogen Full length human recombinant protein of human UCHL1 (NP_004172) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7345
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.