Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGPEP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565936 Supplier Thermo Scientific Supplier No. PA565936
Add to Cart
Add to Cart

PGPEP1 Polyclonal Antibody

PGPEP1 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen PGPEP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PGPEP1
Gene Accession No. Q9NXJ5
Gene Alias 2810003H13Rik; 5-oxoprolyl-peptidase; Gm19458; PAPI; PAP-I; Pcp; PGI; pGlu-peptidase I; PGP; PGPEP1; PGPI; PGP-I; putative pyroglutamyl-peptidase I; pyroglutamyl aminopeptidase I; Pyroglutamyl-peptidase 1; pyroglutamyl-peptidase I; Pyrrolidone-carboxylate peptidase
Gene Symbols PGPEP1
Host Species Rabbit
Immunogen Recombinant Human PGPEP1. Recombinant protein control fragment (Product #RP-106618).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54858
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.