Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGPEP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114267 Supplier Invitrogen™ Supplier No. PA5114267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KHSPQLVVHV GVSGMATTVT LEKCGHNKGY KGLDNCRFCP GSQCCVEDGP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.

Pyroglutamyl-peptidase I (PGPEP1) is a gene located on chromosome 19q13.33, encoding an enzyme involved in the removal of pyroglutamyl residues from the N-terminus of peptides and proteins. This enzymatic activity is fundamental in protein processing and metabolism, ensuring proper protein maturation and degradation. PGPEP1 is expressed in multiple tissues, including the liver and kidney, and plays a role in the regulation of biological processes by modulating the activity and stability of pyroglutamyl-containing bioactive peptides. The enzyme's function is crucial for maintaining cellular homeostasis and can influence physiological pathways such as hormone regulation and immune responses. Dysregulation or mutations in PGPEP1 may impact peptide function and have potential implications for various diseases, although the specific clinical significance is still being researched. Insights into PGPEP1's function are contributing to the understanding of protease-related disorders and therapeutic strategies targeting peptide metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGPEP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PGPEP1
Gene Accession No. Q9NXJ5
Gene Alias 2810003H13Rik; 5-oxoprolyl-peptidase; Gm19458; PAPI; PAP-I; Pcp; PGI; pGlu-peptidase I; PGP; PGPEP1; PGPI; PGP-I; putative pyroglutamyl-peptidase I; pyroglutamyl aminopeptidase I; Pyroglutamyl-peptidase 1; pyroglutamyl-peptidase I; Pyrrolidone-carboxylate peptidase
Gene Symbols PGPEP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Human PGPEP1 (aa 64-113).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54858
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.