Learn More
Invitrogen™ PGRMC1 Monoclonal Antibody (6F9A1)

Description
MA5-31840 has been tested in indirect ELISA.
Specifications
Specifications
| Antigen | PGRMC1 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6F9A1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | PGRMC1 |
| Gene Accession No. | O00264 |
| Gene Alias | 25dx; 25-Dx; AA415812; Acidic 25 kDa protein; Dap1; HGNC:16090; HPR6.6; IZA; Lewi; membrane steroid binding protein; membrane-associated progesterone receptor component 1; membrane-bound progesterone receptor; MPR; Pgrmc; Pgrmc1; PPMR; progesterone binding protein; progesterone receptor membrane component 1; steroid membrane binding protein; VEMA; Ventral midline antigen |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.