Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGRMC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518496
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518496 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518496 Supplier Invitrogen™ Supplier No. PA518496
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGRMC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PGRMC1
Gene Accession No. O00264, O55022
Gene Alias 25dx; 25-Dx; AA415812; Acidic 25 kDa protein; Dap1; HGNC:16090; HPR6.6; IZA; Lewi; membrane steroid binding protein; membrane-associated progesterone receptor component 1; membrane-bound progesterone receptor; MPR; Pgrmc; Pgrmc1; PPMR; progesterone binding protein; progesterone receptor membrane component 1; steroid membrane binding protein; VEMA; Ventral midline antigen
Gene Symbols PGRMC1
Host Species Goat
Immunogen Synthetic peptide sequence (EPKDESARKND) corresponding to the C-terminus amino acids of PGRMC1 (aa 185-195).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10857, 53328
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.