Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGRMC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542919 Supplier Thermo Scientific Supplier No. PA542919
Add to Cart
Add to Cart

PGRMC1 Polyclonal Antibody

PGRMC1 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen PGRMC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PGRMC1
Gene Accession No. O00264
Gene Alias 25dx; 25-Dx; AA415812; Acidic 25 kDa protein; Dap1; HGNC:16090; HPR6.6; IZA; Lewi; membrane steroid binding protein; membrane-associated progesterone receptor component 1; membrane-bound progesterone receptor; MPR; Pgrmc; Pgrmc1; PPMR; progesterone binding protein; progesterone receptor membrane component 1; steroid membrane binding protein; VEMA; Ventral midline antigen
Gene Symbols PGRMC1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PGRMC1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10857
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.