Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGRMC2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149716AF594
Description
PGRMC2 Polyclonal antibody specifically detects PGRMC2 in Human samples. It is validated for Western BlotSpecifications
PGRMC2 | |
Polyclonal | |
Western Blot | |
DG6progesterone membrane binding protein, membrane-associated progesterone receptor component 2, PMBPProgesterone membrane-binding protein, progesterone receptor membrane component 2, Steroid receptor protein DG6 | |
The immunogen for this product maps to a region between residue 1 and 50 of human Progesterone Receptor Membrane Component 2 using the numbering given in entry NP_006311.1 (GeneID 10424). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10424 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction