Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGRMC2 Polyclonal Antibody

Catalog No. PIPA5110103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110103 Supplier Invitrogen™ Supplier No. PA5110103

Rabbit Polyclonal Antibody

Immunogen sequence: MAAGDGDVKL GTLGSGSESS NDGGSESPGD AGAAAEGGGW AAAALALLTG.

PGRMC2 (progesterone receptor membrane component 2), also known as DG6 (steroid receptor protein DG6) or PMBP (progesterone membrane-binding protein), is a single pass membrane protein belonging to the cytochrome b5 family (MAPR (membrane associated progesterone receptor) subfamily). Expressed in sperm, PGRMC2 is believed to function as a steroid receptor and may participate in the progesterone-dependent sperm acrosome reaction. PGRMC2 shares approximately 50% overall sequence identity with its close relative PGRMC1. The loss of the gene encoding PGRMC2 is associated with metastasis in uterine endocervical adenocarcinomas, implicating a potential role of PGRMC2 in the suppression of metastasis of endocervical adenocarcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGRMC2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Pgrmc2
Gene Accession No. O15173, Q80UU9
Gene Alias 4631434O19Rik; 5730409G06Rik; DG6; Membrane-associated progesterone receptor component 2; Pgrmc2; PMBP; progesterone membrane binding protein; Progesterone membrane-binding protein; progesterone receptor membrane component 2; steroid receptor protein DG6
Gene Symbols Pgrmc2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-50 of human PGRMC2 (NP_0063113).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10424, 70804
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.