Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGRP-1a Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598055
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598055 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598055 Supplier Invitrogen™ Supplier No. PA598055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PGRP-1a Polyclonal Antibody for Western Blot, ELISA

Primary immune recognition is based on structures common among invading pathogens. Bacterial surface molecules, such as lipopolysaccharide (LPS) and peptidoglycan (PGN), are known to elicit immune reactions ranging from cytokine release to fever. Recently, a family of proteins called peptidoglycan recognition protein (PGRP) has been identified in mouse and human that binds to peptidoglycans expressed on Gram-positive bacteria. Peptidoglycan (PGN) is an essential cell wall component of virtually all bacteria (1, 2) and, thus, it is an excellent target for recognition by the eukaryotic innate immune system. The PGRPs (PGRP-L, PGRP-S, PGRP-Ia, and PGRP-I beta) define a new family of human pattern recognition molecules (3). PGRP-L is primarily expressed in the liver. Although liver is not considered a primary immune organ, liver participates in host defenses by producing acute phase proteins (by hepatocytes) in response to infections and by clearing microorganisms from blood (4). PGRP-1a mRNA is highly expressed in esophagus, tonsils and thymus and to lesser extent in the stomach, rectum, and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGRP-1a
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PGLYRP3
Gene Accession No. Q96LB9
Gene Alias Gm420; MGC149197; Peptidoglycan recognition protein 3; peptidoglycan recognition protein I alpha; peptidoglycan recognition protein I-alpha; Peptidoglycan recognition protein intermediate alpha; PGLYRP3; PGLYRPIalpha; PGRPIA; PGRP-Ialpha; PGRP-I-alpha
Gene Symbols PGLYRP3
Host Species Rabbit
Immunogen Recombinant Human Peptidoglycan recognition protein 3 protein (90-222AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114771
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.