Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PGRP-L Monoclonal Antibody (439728)
GREENER_CHOICE

Catalog No. PIMA524166
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524166 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524166 Supplier Invitrogen™ Supplier No. MA524166
Add to Cart
Add to Cart

Rat Monoclonal Antibody

In Western blots, no cross-reactivity with recombinant mouse (rm) PGRP-S and rmPGRP-1 beta is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Peptidoglycan recognition proteins, such as PGRPL, are part of the innate immune system and recognize peptidoglycan, a ubiquitous component of bacterial cell walls.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGRP-L
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 439728
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PGLYRP2
Gene Accession No. Q8VCS0
Gene Alias C730002N09Rik; HMFT0141; N-acetylmuramoyl-L-alanine amidase; Peptidoglycan recognition protein 2; peptidoglycan recognition protein L; peptidoglycan recognition protein long; PGLYRP2; Pglyrpl; PGRPL; PGRP-L; TagL; tagL-alpha; tagl-beta; TAGL-like; UNQ3103/PRO10102
Gene Symbols PGLYRP2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PGRP-L Ser23-Asn530.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57757
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.