Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PGS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112945
Description
PGS1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
PGS1 functions in the biosynthesis of the anionic phospholipids phosphatidylglycerol and cardiolipin.
Specifications
| PGS1 | |
| Polyclonal | |
| Unconjugated | |
| PGS1 | |
| 2610019F11Rik; 4933424M23Rik; CDP-diacylglycerol--glycerol-3-phosphate 3-phosphatidyltransferase, mitochondrial; PGP synthase 1; PGS1; Phosphatidylglycerophosphate synthase 1; RGD1305052; Saf; silencer-associated factor | |
| Rabbit | |
| Protein G | |
| RUO | |
| 74451, 9489 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q32NB8, Q8BHF7 | |
| PGS1 | |
| Recombinant Human CDP-diacylglycerol--glycerol-3-phosphate 3-phosphatidyltransferase, mitochondrial protein (110-400AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction