Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pH 10 Reference Standard solution Buffer, Color coded blue, Macron Fine Chemicals™

Catalog No. MK0099500
Click to view available options
Quantity:
500 mL
Packaging:
Poly Container

Specifications

Chemical Name or Material Buffer, pH 10 Reference standard solution
Color Blue
pH 9.99 to 10.01 (5% soln. at 25°C)
Physical Form Liquid
Quantity 500 mL
Packaging Poly Container
Traceability to NIST Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.