Learn More
Tocris Bioscience™ PH 797804

Description
PH 797804 is the (-)-atropisomer, which is 100-fold more potent than its (+)-atropisomer.
Specifications
Specifications
| Chemical Name or Material | (-)-3-[3-Bromo-4-[(2,4-difluorophenyl)methoxy]-6-methyl-2-oxo-1(2H)-pyridinyl]-N,4-dimethylbenzamide |
| CAS | 1358027-80-1 |
| Quantity | 10 mg |
| Target | p38 Inhibitors |
| Molecular Formula | C22H19BrF2N2O3 |
| Formula Weight | 477.3 |
| Purity | >98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.