Learn More
Oakton™ pH Electrode, Direct Connection
Catalog No. 13300139
Click to view available options
Connector Types:
BNC
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with samples.
  • Compatible with BNC Connections
Specifications
Connector Types BNC
Junction Single
Material Epoxy
pH Range 0 to 12
Type Direct Connection
Diameter (Metric) 9 mm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.