Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ pH Paper Strips
Shop All Fisherbrand Products

Click to view available options
pH Range:
0 to 14
0 to 3
10 to 12
3 to 5.5
6 to 8
8 to 9.5
Type:
pH Test Paper
pH Test Paper Set
Description
- Ideal for educational use
- Low-cost paper strips are easy to pass around the classroom
- Easy-to-usedash (em)simply dip strip in solution to be tested and match to color chart immediately
Order Info
Box of 10 vials, 100 low-cost paper strips per vial.
Specifications
Specifications
pH Range | 3 to 5.5 |
For Use With (Application) | For classroom experiments or demonstrations |
Quantity | 100 Tests/Vial, 10 Vials/Cs. |
Type | pH Test Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction