Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ pH and pH/ORP Sensors for Model 556 Multi-Probe System

Catalog No. p-193053
Click to view available options
Type:
pH Probe
pH and pH/ORP probe

Measure pH and pH & ORP/Redox quickly and easily

  • User-replaceble sensor allows for quick sensor replacements if needed
  • No tools necessary

TRUSTED_SUSTAINABILITY

Specifications

Type pH and pH/ORP probe
For Use With (Application) Aquaculture, agriculture, chemistry, food and beverage, wine-making and water-testing, for scientific applications
For Use With (Equipment) YSI model 556 handheld multi probe meter
Includes Storage bottle with storage solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.