Learn More
Promega pHAb Thiol Reactive Dye
Catalog No. PRG9835 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
250 μg
4 x 250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9835 4 x 250 μg
PRG9831 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG9835 Supplier Promega Supplier No. G9835
pHAb Amine Reactive Dye exhibits very low fluorescence at pH > 7 and much higher fluorescence as the pH of the solution becomes acidic. pHAb Dyes are designed specifically for antibody labeling.
  • Measure Antibody Internalization in Real Time
  • Compatible with 96-well, plate-based assay
  • High signal-to-background ratios
  • On-Bead conjugation
Specifications
Quantity 4 x 250 μg
Content And Storage -30° to -10°C for 1 month and <-65°C for long-term storage