Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHACTR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598056
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598056 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598056 Supplier Invitrogen™ Supplier No. PA598056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHACTR1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

PHACTR1 plays a role in cell motility and in the formation of tubules by endothelial cells. PHACTR1 also binds actin monomers (G actin) and plays a role in the reorganization of the actin cytoskeleton and in formation of actin stress fibers. Further, PHACTR1 regulates PPP1CA activity and is required for normal cell survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHACTR1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Phactr1
Gene Accession No. P62024, Q2M3X8, Q9C0D0
Gene Alias 9630030F18Rik; dJ257A7.2; KIAA1733; LOC100051555; MGC126575; MGC126577; phactr1; Phosphatase and actin regulator 1; RPEL; RPEL repeat containing 1; Rpel1
Gene Symbols Phactr1
Host Species Rabbit
Immunogen Recombinant Human Phosphatase and actin regulator 1 protein (263-387AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218194, 221692, 306844
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.