Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHACTR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PHACTR1 |
---|---|
Concentration | 0.53 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4078
![]() ![]() |
Proteintech Group Inc
234461AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PHACTR1 plays a role in cell motility and in the formation of tubules by endothelial cells. PHACTR1 also binds actin monomers (G actin) and plays a role in the reorganization of the actin cytoskeleton and in formation of actin stress fibers. Further, PHACTR1 regulates PPP1CA activity and is required for normal cell survival.Specifications
PHACTR1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P62024, Q2M3X8, Q9C0D0 | |
218194, 221692, 306844 | |
PHACTR1 Fusion Protein Ag20125 | |
Primary | |
-20°C | |
PHACTR1 |
0.53 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA1733, PHACTR1, RPEL, RPEL1 | |
Phactr1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title