Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHACTR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565682
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565682 Supplier Invitrogen™ Supplier No. PA565682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLKQTTSALEK KMAGRQGREE LIKKGLLEMM EQDAESKTCN PDGGPRSVQS EPPTPKSETL TSEDAQPGS Highest antigen sequence identity to the following orthologs - mouse 79%, rat 67%.

The protein encoded by this gene is associated with the nuclear scaffold in proliferating cells. It was found to bind to the catalytic subunit of protein phosphatase-1 (PP1) and inhibit PP1 activity, suggesting that this protein may function as a regulatory subunit of PP1. Alternative splicing at this locus results in several transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHACTR3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHACTR3
Gene Accession No. Q96KR7
Gene Alias 1500003N10Rik; 4930415A02Rik; C20orf101; H17739; LOW QUALITY PROTEIN: phosphatase and actin regulator 3; mKIAA4224; Phactr3; Phosphatase and actin regulator 3; PPP1R123; protein phosphatase 1, regulatory subunit 123; scaffold-associated PP1-inhibiting protein; Scapin1; Scapinin
Gene Symbols PHACTR3
Host Species Rabbit
Immunogen Recombinant Human PHACTR3. Recombinant protein control fragment (Product #RP-106354).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.