Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHACTR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118161
This item is not returnable.
View return policy
Description
PHACTR3 Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is associated with the nuclear scaffold in proliferating cells. It was found to bind to the catalytic subunit of protein phosphatase-1 (PP1) and inhibit PP1 activity, suggesting that this protein may function as a regulatory subunit of PP1. Alternative splicing at this locus results in several transcript variants encoding different isoforms.
Specifications
| PHACTR3 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q96KR7 | |
| PHACTR3 | |
| E. coli-derived Human PHACTR3 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| PHACTR3 | |
| 1500003N10Rik; 4930415A02Rik; C20orf101; H17739; LOW QUALITY PROTEIN: phosphatase and actin regulator 3; mKIAA4224; Phactr3; Phosphatase and actin regulator 3; PPP1R123; protein phosphatase 1, regulatory subunit 123; scaffold-associated PP1-inhibiting protein; Scapin1; Scapinin | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 116154 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction