Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHACTR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531616
Description
Recommended positive controls: THP-1. Predicted reactivity: Mouse (82%), Rat (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is associated with the nuclear scaffold in proliferating cells. It was found to bind to the catalytic subunit of protein phosphatase-1 (PP1) and inhibit PP1 activity, suggesting that this protein may function as a regulatory subunit of PP1. Alternative splicing at this locus results in several transcript variants encoding different isoforms.
Specifications
PHACTR3 | |
Polyclonal | |
Unconjugated | |
PHACTR3 | |
1500003N10Rik; 4930415A02Rik; C20orf101; H17739; LOW QUALITY PROTEIN: phosphatase and actin regulator 3; mKIAA4224; Phactr3; Phosphatase and actin regulator 3; PPP1R123; protein phosphatase 1, regulatory subunit 123; scaffold-associated PP1-inhibiting protein; Scapin1; Scapinin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.96 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q96KR7 | |
PHACTR3 | |
Recombinant fragment corresponding to a region within amino acids 85 and 395 of Human PHACTR3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction