Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHACTR4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101747
Description
Antibody detects endogenous levels of total PHACTR4.
This gene encodes a member of the phosphatase and actin regulator (PHACTR) family. Other PHACTR family members have been shown to inhibit protein phosphatase 1 (PP1) activity, and the homolog of this gene in the mouse has been shown to interact with actin and PP1. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
PHACTR4 | |
Polyclonal | |
Unconjugated | |
PHACTR4 | |
3110001B12Rik; AI527228; AW495572; C330013F19Rik; humdy; Kiaa4120; mKIAA4120; N28169; PHACTR4; Phosphatase and actin regulator 4; PPP1R124; PRO2963; Protein Humpty dumpty; protein phosphatase 1, regulatory subunit 124 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
65979, 682920 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8IZ21 | |
PHACTR4 | |
A synthesized peptide derived from human PHACTR4(Accession Q8IZ21), corresponding to amino acid residues P362-L412. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction